Gene Transcripts
- Gene Symbol
- Olfr1241
- Entrez Gene ID
- 258447
- Gene Name
- olfactory receptor 1241
- Gene Aliases
- MOR231-14
- Chromosome Location
- Chr. 2: 89482189 - 89483133 on Build GRCm38
- UniGene ID
- Mm.271375
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146455.1 |
NP_666666.1
|
1 |
459 |
|
View Full Product Details