Gene Transcripts

Gene Symbol
Olfr716
Entrez Gene ID
258597
Gene Name
olfactory receptor 716
Gene Aliases
MOR260-2
Chromosome Location
Chr. 7: 107147318 - 107148262 on Build GRCm38
UniGene ID
Mm.371710
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146604.1 NP_666815.1 1 742
GenBank mRNA BC120510.1 - 1 834

View Full Product Details