Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159331.1 | NP_001152803.1 | 4 | 419 | |
NM_026366.2 | NP_080642.1 | 4 | 419 | ||
GenBank mRNA | AK012019.1 | - | 4 | 359 | |
AK145617.1 | - | 4 | 371 | ||
AK160298.1 | - | 4 | 381 | ||
AY456393.1 | - | 4 | 419 | ||
AY536887.1 | - | 4 | 351 | ||
AY583759.1 | - | 4 | 463 | ||
BC098330.1 | - | 4 | 361 | 30548526 | |
BC116393.1 | - | 4 | 389 | 40101877 | |
BC116394.1 | - | 4 | 389 | 40101881 |