Gene Transcripts
- Gene Symbol
- Ctc1
- Entrez Gene ID
- 68964
- Gene Name
- CTS telomere maintenance complex component 1
- Gene Aliases
- 1500010J02Rik, AAF-132, AAF132
- Chromosome Location
- Chr. 11: 69015904 - 69036473 on Build GRCm38
- UniGene ID
- Mm.285785
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001013256.2 |
NP_001013274.2
|
12, 13 |
2280 |
|
NM_001143790.1 |
NP_001137262.1
|
12, 13 |
2280 |
|
NM_001281464.1 |
NP_001268393.1
|
12, 13 |
2151 |
|
NM_001281465.1 |
NP_001268394.1
|
12, 13 |
2264 |
|
GenBank mRNA |
AK038651.1 |
- |
12, 13 |
2067 |
|
AK044055.1 |
- |
12, 13 |
2059 |
|
AK145919.1 |
- |
12, 13 |
2095 |
|
AK165906.1 |
- |
12, 13 |
2080 |
|
BC014687.1 |
- |
3, 4 |
451 |
|
BC060629.1 |
- |
8, 9 |
1330 |
6847619 |
BC066078.1 |
- |
12, 13 |
2095 |
|
BC072625.1 |
- |
12, 13 |
2108 |
|
BC147495.1 |
- |
12, 13 |
2078 |
9056325 |
BC147497.1 |
- |
12, 13 |
2078 |
9056354 |
View Full Product Details