Gene Transcripts
- Gene Symbol
- Gm3519
- Entrez Gene ID
- 100041805
- Gene Name
- predicted gene 3519
- Gene Aliases
- Chromosome Location
- Chr. 5: 52440680 - 52442952 on Build GRCm38
- UniGene ID
- Mm.417070
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK139157.1 |
- |
2 |
584 |
|
View Full Product Details