Gene Transcripts

Gene Symbol
Mipep
Entrez Gene ID
70478
Gene Name
mitochondrial intermediate peptidase
Gene Aliases
5730405E07Rik, MIP
Chromosome Location
Chr. 14: 60784562 - 60905481 on Build GRCm38
UniGene ID
Mm.274650
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK044460.1 - 16 2895
BC059092.1 - 2 599
BC144798.1 - 20 3282
BC145711.1 - 20 3282 40131014

View Full Product Details