Gene Transcripts

Gene Symbol
Hist1h1t
Entrez Gene ID
107970
Gene Name
histone cluster 1, H1t
Gene Aliases
H1.6, H1ft, H1t
Chromosome Location
Chr. 13: 23695811 - 23696545 on Build GRCm38
UniGene ID
Mm.8048
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010377.3 NP_034507.2 1 575
GenBank mRNA BC119305.1 - 1 545
BC119307.1 - 1 545 8734056
BV210386.1 - 1 409

View Full Product Details