Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039155.1 | NP_001034244.1 | 8 | 2589 | |
NM_001039156.1 | NP_001034245.1 | 8 | 2589 | ||
NM_138579.4 | NP_613045.3 | 8 | 2589 | ||
GenBank mRNA | AK155197.1 | - | 1 | 292 | |
BC094366.1 | - | 1 | 1246 | ||
DQ228000.1 | - | 8 | 2589 | ||
DQ228001.2 | - | 8 | 2589 | ||
DQ228002.2 | - | 8 | 2589 | ||
DQ278602.1 | - | 7 | 2208 |