Gene Transcripts

Gene Symbol
Serpina6
Entrez Gene ID
12401
Gene Name
serine (or cysteine) peptidase inhibitor, clade A, member 6
Gene Aliases
AI265318, AV104445, Cbg
Chromosome Location
Chr. 12: 103646632 - 103657071 on Build GRCm38
UniGene ID
Mm.290079
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007618.2 NP_031644.1 3 803
GenBank mRNA AK145842.1 - 3 835
BC013632.1 - 3 822 4194164
X70533.1 - 3 803

View Full Product Details