Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010057.2 | NP_034187.1 | 3 | 818 | |
GenBank mRNA | AB221684.1 | - | 3 | 406 | |
AF022078.1 | - | 3 | 355 | ||
BC114342.1 | - | 3 | 402 | ||
EF535989.1 | - | 3 | 818 | ||
EF535990.1 | - | 4 | 673 | ||
U67841.1 | - | 2 | 251 |