Gene Transcripts
- Gene Symbol
- Hspb1
- Entrez Gene ID
- 15507
- Gene Name
- heat shock protein 1
- Gene Aliases
- 27kDa, Hsp25
- Chromosome Location
- Chr. 5: 135887919 - 135889563 on Build GRCm38
- UniGene ID
- Mm.13849
- Species
- Mus musculus
- Gene Symbol
- Gm9817
- Entrez Gene ID
- 100303734
- Gene Name
- predicted gene 9817
- Gene Aliases
- ENSMUSG00000047061
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.471748
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK019498.1 |
- |
1 |
100 |
|
View Full Product Details