Gene Transcripts

Gene Symbol
Hspg2
Entrez Gene ID
15530
Gene Name
perlecan (heparan sulfate proteoglycan 2)
Gene Aliases
AI852380, Pcn, Plc, per
Chromosome Location
Chr. 4: 137468764 - 137570630 on Build GRCm38
UniGene ID
Mm.273662
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008305.3 NP_032331.2 25, 26 3359
GenBank mRNA AK147294.1 - 25, 26 3360
AK147396.1 - 25, 26 3360

View Full Product Details