Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010745.2 | NP_034875.1 | 4 | 459 | |
GenBank mRNA | AB007599.1 | - | 4 | 459 | |
AK027929.1 | - | 4 | 448 | ||
BC065783.1 | - | 4 | 364 | ||
BC096414.1 | - | 4 | 427 | 849991 | |
BE200280.1 | - | 2 | 535 |