Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081981.2 | NP_001075450.1 | 10 | 2428 | |
NM_001081982.2 | NP_001075451.1 | 9 | 1605 | ||
NM_001297601.1 | NP_001284530.1 | 8 | 2154 | ||
NM_010906.3 | NP_035036.1 | 8 | 1482 | ||
GenBank mRNA | AK080940.1 | - | 2 | 154 | |
AK158568.1 | - | 8 | 2157 | ||
BC003766.1 | - | 9 | 1327 | 3491917 | |
BC090626.1 | - | 10 | 1455 | ||
U57636.1 | - | 7 | 1159 | ||
Y07688.1 | - | 9 | 1607 | ||
Y07689.1 | - | 8 | 1286 |