Gene Transcripts

Gene Symbol
Zfp871
Entrez Gene ID
208292
Gene Name
zinc finger protein 871
Gene Aliases
4732483N19, 9030612M13Rik, AI118577, AI265322, mKIAA3006
Chromosome Location
Chr. 17: 32765496 - 32788289 on Build GRCm38
UniGene ID
Mm.38813
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172458.3 NP_766046.3 4 1502
GenBank mRNA AK029041.1 - 4 1490
AK122579.1 - 4 1465
BC043671.1 - 4 1617 5040597

View Full Product Details