Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001122754.2 | NP_001116226.1 | 11 | 1058 | |
NM_001289494.1 | NP_001276423.1 | 10 | 992 | ||
NM_001289495.1 | NP_001276424.1 | 11 | 1075 | ||
NM_013698.3 | NP_038726.2 | 10 | 1142 | ||
GenBank mRNA | AK138069.1 | - | 11 | 1055 | |
AK138786.1 | - | 11 | 1072 | ||
BC120552.1 | - | 10 | 1045 | 40129475 | |
BC137710.1 | - | 10 | 1047 | 8860729 | |
D43963.1 | - | 10 | 1142 | ||
D83201.1 | - | 10 | 1082 | ||
D83202.1 | - | 10 | 1245 | ||
U16145.1 | - | 11 | 1023 | ||
U19607.1 | - | 11 | 1058 |