Gene Transcripts

Gene Symbol
Olfr574
Entrez Gene ID
258357
Gene Name
olfactory receptor 574
Gene Aliases
MOR14-9
Chromosome Location
Chr. 7: 102946411 - 102949507 on Build GRCm38
UniGene ID
Mm.377454
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146360.2 NP_666472.2 1 32
GenBank mRNA BC127050.1 - 1 30

View Full Product Details