Gene Transcripts
- Gene Symbol
- Olfr918
- Entrez Gene ID
- 258372
- Gene Name
- olfactory receptor 918
- Gene Aliases
- MOR164-3
- Chromosome Location
- Chr. 9: 38672504 - 38673481 on Build GRCm38
- UniGene ID
- Mm.377462
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146375.2 |
NP_666487.2
|
1 |
39 |
|
View Full Product Details