Gene Transcripts

Gene Symbol
Ccdc42b
Entrez Gene ID
546886
Gene Name
coiled-coil domain containing 42B
Gene Aliases
Gm5988
Chromosome Location
Chr. 5: 120628335 - 120634235 on Build GRCm38
UniGene ID
Mm.464847
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195094.1 NP_001182023.1 5 573

View Full Product Details