Gene Transcripts

Gene Symbol
Gm11437
Entrez Gene ID
628813
Gene Name
predicted gene 11437
Gene Aliases
AI463690, OTTMUSG00000000997
Chromosome Location
Chr. 11: 84148361 - 84167476 on Build GRCm38
UniGene ID
Mm.390389
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037932.2 NP_001033021.2 6 799
GenBank mRNA AJ844653.1 - 6 799
BC147775.1 - 6 791 9056710
BC147791.1 - 6 791 9056714

View Full Product Details