Gene Transcripts

Gene Symbol
Dip2a
Entrez Gene ID
64451
Gene Name
DIP2 disco-interacting protein 2 homolog A (Drosophila)
Gene Aliases
4931420H10Rik, AI426328, Dip2, Kiaa0184-hp, mKIAA0184
Chromosome Location
Chr. 10: 76263049 - 76345301 on Build GRCm38
UniGene ID
Mm.33183
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081419.2 NP_001074888.2 3 275
GenBank mRNA AK052899.1 - 3 217

View Full Product Details