Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025553.4 | NP_079829.1 | 3 | 334 | |
GenBank mRNA | AB049639.1 | - | 3 | 312 | |
AK002797.1 | - | 3 | 289 | ||
AK010329.1 | - | 3 | 326 | ||
AK167247.1 | - | 3 | 302 | ||
BC046750.1 | - | 3 | 287 | 5692108 |