Gene Transcripts

Gene Symbol
Mis18a
Entrez Gene ID
66578
Gene Name
MIS18 kinetochore protein homolog A (S. pombe)
Gene Aliases
2610039C10Rik, 2810018N07Rik
Chromosome Location
Chr. 16: 90719312 - 90727371 on Build GRCm38
UniGene ID
Mm.180363
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025642.1 NP_079918.1 1 198
GenBank mRNA AK012533.1 - 1 199
AK014084.1 - 1 167
AK163310.1 - 1 232
BC079900.1 - 1 210 30602551

View Full Product Details