Gene Transcripts

Gene Symbol
Gm8787
Entrez Gene ID
667736
Gene Name
predicted gene 8787
Gene Aliases
EG667736
Chromosome Location
Chr. X: 79330513 - 79358068 on Build GRCm38
UniGene ID
Mm.484619
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099310.1 NP_001092780.1 4 543
GenBank mRNA BY735733.1 - 4 543

View Full Product Details