Gene Transcripts

Gene Symbol
Tex43
Entrez Gene ID
67343
Gene Name
testis expressed 43
Gene Aliases
1700065I17Rik, Tseg7
Chromosome Location
Chr. 18: 56587613 - 56594783 on Build GRCm38
UniGene ID
Mm.87057
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026099.3 NP_080375.2 3 322
GenBank mRNA AK006893.1 - 3 323
AK018985.1 - 3 322
BC099538.1 - 3 319 30916077
FJ211594.1 - 3 306

View Full Product Details