Gene Transcripts

Gene Symbol
Sympk
Entrez Gene ID
68188
Gene Name
symplekin
Gene Aliases
1500016F02Rik, 4632415H16Rik, AA125406, AI449890, SPK, SYM
Chromosome Location
Chr. 7: 19024352 - 19054623 on Build GRCm38
UniGene ID
Mm.130902
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026605.2 NP_080881.2 22 2989
GenBank mRNA AK019497.1 - 17 2466
AK146894.1 - 22 2970
AK155093.1 - 22 2990
AK155375.1 - 22 2944
AK165591.1 - 22 2968
AK168541.1 - 22 2970
AK168757.1 - 22 2970
AK172040.1 - 23 3018
BC049852.1 - 22 2934 4163760

View Full Product Details