Gene Transcripts

Gene Symbol
1810011H11Rik
Entrez Gene ID
69069
Gene Name
RIKEN cDNA 1810011H11 gene
Gene Aliases
4733401I05Rik
Chromosome Location
Chr. 14: 32785885 - 32817969 on Build GRCm38
UniGene ID
Mm.261609
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163616.1 NP_001157088.1 3 212
GenBank mRNA AK007434.1 - 3 213

View Full Product Details