Gene Transcripts

Gene Symbol
1700001J03Rik
Entrez Gene ID
69282
Gene Name
RIKEN cDNA 1700001J03 gene
Gene Aliases
Chromosome Location
Chr. 5: 146182450 - 146185304 on Build GRCm38
UniGene ID
Mm.373573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008547.1 NP_001008547.1 3, 4 414
GenBank mRNA AK005597.1 - 3, 4 410
BC048528.1 - 3, 4 414 6774214

View Full Product Details