Gene Transcripts

Gene Symbol
1700001J03Rik
Entrez Gene ID
69282
Gene Name
RIKEN cDNA 1700001J03 gene
Gene Aliases
Chromosome Location
Chr. 5: 146182450 - 146185304 on Build GRCm38
UniGene ID
Mm.373573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008547.1 NP_001008547.1 4 492
GenBank mRNA BC048528.1 - 4 492

View Full Product Details