Gene Transcripts

Gene Symbol
Kif1bp
Entrez Gene ID
72320
Gene Name
KIF1 binding protein
Gene Aliases
0710007C18Rik, 2510003E04Rik, KBP, mKIAA1279
Chromosome Location
Chr. 10: 62558470 - 62578464 on Build GRCm38
UniGene ID
Mm.258955
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028197.2 NP_082473.2 5 885
GenBank mRNA AK035659.1 - 5 873
AK129320.1 - 5 870
AK141324.1 - 5 893
AK149629.1 - 5 822
AK158730.1 - 6 620
AK161007.1 - 871
AK206480.1 - 2 59
AK214549.1 - 2 59
AK216082.1 - 2 137
BC038828.1 - 1 37
BC046485.1 - 5 856 5350638
BC055307.1 - 5 839 6493547
BC090974.1 - 5 839 6774777

View Full Product Details