Gene Transcripts

Gene Symbol
6030468B19Rik
Entrez Gene ID
77727
Gene Name
RIKEN cDNA 6030468B19 gene
Gene Aliases
Chromosome Location
Chr. 11: 117797627 - 117807308 on Build GRCm38
UniGene ID
Mm.159981
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029964.1 NP_084240.1 3 434
GenBank mRNA AK020081.1 - 3 359

View Full Product Details