Gene Transcripts

Gene Symbol
Slc38a4
Entrez Gene ID
170573
Gene Name
solute carrier family 38, member 4
Gene Aliases
Ata3
Chromosome Location
Chr. 7: 138459576 - 138512952 on Build Rnor_6.0
UniGene ID
Rn.17990
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130748.1 NP_570104.1 3 287
XM_008765663.1 XP_008763885.1 3 282
GenBank mRNA AF295535.1 - 3 287
BC097292.1 - 4 215 7462761

View Full Product Details