Gene Transcripts

Gene Symbol
Necab2
Entrez Gene ID
170928
Gene Name
N-terminal EF-hand calcium binding protein 2
Gene Aliases
Efcbp2
Chromosome Location
Chr. 19: 52086325 - 52112633 on Build Rnor_6.0
UniGene ID
Rn.62703
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133415.1 NP_596906.1 11 957
XM_006255703.2 XP_006255765.1 9 2023
GenBank mRNA AF193757.1 - 11 957

View Full Product Details