Gene Transcripts

Gene Symbol
Optn
Entrez Gene ID
246294
Gene Name
optineurin
Gene Aliases
Fip2
Chromosome Location
Chr. 17: 77167701 - 77218374 on Build Rnor_6.0
UniGene ID
Rn.63435
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145081.3 NP_659549.2 2 269
GenBank mRNA AB069907.3 - 2 269

View Full Product Details