Gene Transcripts

Gene Symbol
Myo1e
Entrez Gene ID
25484
Gene Name
myosin IE
Gene Aliases
MYR5, Myr3
Chromosome Location
Chr. 8: 76644715 - 76840240 on Build Rnor_6.0
UniGene ID
Rn.8737
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173101.1 NP_775124.1 12, 13 1649
GenBank mRNA X74815.1 - 12, 13 1649

View Full Product Details