Gene Transcripts

Gene Symbol
Bmp3
Entrez Gene ID
25667
Gene Name
bone morphogenetic protein 3
Gene Aliases
PBMP3
Chromosome Location
Chr. 14: 12362912 - 12387572 on Build Rnor_6.0
UniGene ID
Rn.208979
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017105.1 NP_058801.1 1 768
GenBank mRNA D63860.1 - 1 768

View Full Product Details