Gene Transcripts

Gene Symbol
Il2ra
Entrez Gene ID
25704
Gene Name
interleukin 2 receptor, alpha
Gene Aliases
IL2RAC
Chromosome Location
Chr. 17: 70500672 - 70547929 on Build Rnor_6.0
UniGene ID
Rn.9872
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013163.1 NP_037295.1 4 599
GenBank mRNA FQ228242.1 - 631
M55049.1 - 4 599

View Full Product Details