Gene Transcripts

Gene Symbol
Olr1589
Entrez Gene ID
289251
Gene Name
olfactory receptor 1589
Gene Aliases
Chromosome Location
Chr. 13: 92167643 - 92168581 on Build Rnor_6.0
UniGene ID
Rn.142339
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000083.1 NP_001000083.1 1 151

View Full Product Details