Gene Transcripts

Gene Symbol
Olr1600
Entrez Gene ID
289260
Gene Name
olfactory receptor 1600
Gene Aliases
Chromosome Location
Chr. 13: 92376915 - 92377844 on Build Rnor_6.0
UniGene ID
Rn.142330
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000086.1 NP_001000086.1 1 19

View Full Product Details