Gene Transcripts

Gene Symbol
Dntt
Entrez Gene ID
294051
Gene Name
DNA nucleotidylexotransferase
Gene Aliases
Chromosome Location
Chr. 1: 260289626 - 260321174 on Build Rnor_6.0
UniGene ID
Rn.143741
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012461.1 NP_001012479.1 1 160
GenBank mRNA BC089904.1 - 1 160

View Full Product Details