Gene Transcripts

Gene Symbol
Pxmp2
Entrez Gene ID
29533
Gene Name
peroxisomal membrane protein 2
Gene Aliases
T44
Chromosome Location
Chr. 12: 52452354 - 52462655 on Build Rnor_6.0
UniGene ID
Rn.10292
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031587.1 NP_113775.1 4 491
GenBank mRNA AY327502.1 - 3 345
X70223.1 - 4 491

View Full Product Details