Gene Transcripts

Gene Symbol
Csnk2a2
Entrez Gene ID
307641
Gene Name
casein kinase 2, alpha prime polypeptide
Gene Aliases
Chromosome Location
Chr. 19: 9972537 - 10012043 on Build Rnor_6.0
UniGene ID
Rn.24013
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107409.1 NP_001100879.1 9 767
XM_006255082.1 XP_006255144.1 9 767
GenBank mRNA BC168200.1 - 9 783

View Full Product Details