Gene Transcripts

Gene Symbol
Dnajb1
Entrez Gene ID
361384
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 1
Gene Aliases
Chromosome Location
Chr. 19: 24747178 - 24750919 on Build Rnor_6.0
UniGene ID
Rn.128947
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108441.1 NP_001101911.1 4 1062
XM_006255282.2 XP_006255344.1 3 1096
GenBank mRNA BC159430.1 - 3 992

View Full Product Details