Gene Transcripts

Gene Symbol
Sytl2
Entrez Gene ID
361604
Gene Name
synaptotagmin-like 2
Gene Aliases
Chromosome Location
Chr. 1: 154536592 - 154599515 on Build Rnor_6.0
UniGene ID
Rn.7634
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748924.3 XP_003748972.1 7 1560
XM_006229687.2 XP_006229749.1 6 1428
XM_006229694.2 XP_006229756.1 7 1560
XM_006229695.2 XP_006229757.1 7 1561
XM_008759651.1 XP_008757873.1 7 1568
XM_008759652.1 XP_008757874.1 7 1719
XM_008759653.1 XP_008757875.1 7 4107
XM_008759654.1 XP_008757876.1 7 1568
XM_008759655.1 XP_008757877.1 7 1568
XM_008759656.1 XP_008757878.1 7 1568

View Full Product Details