Gene Transcripts

Gene Symbol
Wdr43
Entrez Gene ID
362703
Gene Name
WD repeat domain 43
Gene Aliases
Chromosome Location
Chr. 6: 23433532 - 23473177 on Build Rnor_6.0
UniGene ID
Rn.101883
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037791.2 NP_001032880.2 19 2001
XM_006239770.2 XP_006239832.1 19 2009
GenBank mRNA BC099168.1 - 4 500
BC169117.1 - 20 2198

View Full Product Details