Gene Transcripts

Gene Symbol
Tmx1
Entrez Gene ID
362751
Gene Name
thioredoxin-related transmembrane protein 1
Gene Aliases
Txndc1
Chromosome Location
Chr. 6: 92864123 - 92875141 on Build Rnor_6.0
UniGene ID
Rn.220037
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024800.1 NP_001019971.1 2 261
XM_006240178.2 XP_006240240.1 2 1484
GenBank mRNA BC094308.1 - 2 261

View Full Product Details