Gene Transcripts

Gene Symbol
Nkx2-6
Entrez Gene ID
364418
Gene Name
NK2 homeobox 6
Gene Aliases
Chromosome Location
Chr. 15: 51034620 - 51038715 on Build Rnor_6.0
UniGene ID
Rn.137335
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127653.1 NP_001121125.1 2 104
GenBank mRNA BC160896.1 - 2 104

View Full Product Details