Gene Transcripts

Gene Symbol
Rbmx2
Entrez Gene ID
367930
Gene Name
RNA binding motif protein, X-linked 2
Gene Aliases
RGD1562693
Chromosome Location
Chr. X: 135550931 - 135558440 on Build Rnor_6.0
UniGene ID
Rn.67383
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113786.2 NP_001107258.2 1 92
GenBank mRNA BC158684.1 - 1 92

View Full Product Details