Gene Transcripts

Gene Symbol
Sept3
Entrez Gene ID
56003
Gene Name
septin 3
Gene Aliases
Sep3
Chromosome Location
Chr. 7: 123506535 - 123526542 on Build Rnor_6.0
UniGene ID
Rn.48803
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019375.1 NP_062248.1 3, 4 366
GenBank mRNA AF111179.1 - 3, 4 366

View Full Product Details