Gene Transcripts

Gene Symbol
Igbp1
Entrez Gene ID
58845
Gene Name
immunoglobulin (CD79A) binding protein 1
Gene Aliases
Chromosome Location
Chr. X: 70322764 - 70345005 on Build Rnor_6.0
UniGene ID
Rn.7079
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031624.3 NP_113812.1 1 183
GenBank mRNA AF000577.2 - 1 175
BC068202.1 - 1 187 6921265
FQ212697.1 - 1 192
FQ222671.1 - 1 185

View Full Product Details